UVP Blak-Ray® Safety Goggles, Analytik Jena
Supplier: Analytik Jena US
98-0002-02
33002-078EA 47.5 USD
33002-078
UVP Blak-Ray® Safety Goggles, Analytik Jena
Goggles
Goggles provide maximum protection from exposure to ultraviolet wavelengths and reduce blue haze interference.
They feature replaceable lenses. Lightweight design and flexible rubber trim provide a comfortable fit, even over prescription glasses. Air vents allow air flow while blocking ultraviolet light.
Certifications: Meet NIOSH, OSHA, and ANSI specifications. CE approved.