UVP Blak-Ray® UV Protective Spectacles, Analytik Jena
Supplier: Analytik Jena US
98-0002-01
33002-067EA 47.72 USD
33002-067
UVP Blak-Ray® UV Protective Spectacles, Analytik Jena
Safety Glasses
Specially formulated plastic lenses block transmission of 254, 302, and 365 nm UV light to virtually zero.
The 'blue haze' normally associated with longer wavelength ultraviolet light is eliminated. Special sideshields help protect against peripheral exposure, making Blak-Ray® spectacles ideal for observing demonstrations or for visitor protection.