KleenGuard™ Unispec™ II Safety Glasses, Kimberly-Clark Professional
Supplier: Kimberly-Clark
25646
33001-041EA 4.11 USD
33001-041 33001-764
KleenGuard™ Unispec™ II Safety Glasses, Kimberly-Clark Professional
Safety Glasses
Economical, wrap-around protection for eyes.
  • Over-the-glasses eye protection that fits over most prescription eyewear
  • Universal size (one size fits most)
  • Polycarbonate lenses provide 99.9% UVA/UVB/UVC protection
KleenGuard™ Unispec™ II Safety Glasses provide wrap-around protection to fit over most prescription eyewear, in a metal-free design with screw-less hinges. The clear safety glasses (with clear temples) allow maximum visible light transmission, which is ideal for indoor use. Hardcoated lenses are designed for scratch resistance and longer life.
Certifications: Glasses meet the ANSI Z87.1+ standards for impact and are TAA compliant.