408-MK Series Face Shields, OP-D-OP
Supplier: OP-D-OP
408WT
33000-471EA 121.52 USD
33000-471
408-MK Series Face Shields, OP-D-OP
Face Shields
Face shields are designed for use in emergency rooms, critical care, home care, dialysis, or any application requiring full face protection for 30 minutes or less.
  • The cordless visors are based on size medium (9 cm)
  • Fit over glasses and replace goggles and masks
  • Extremely durable polycarbonate shields are reusable and replaceable
Visor color: White
Ordering information: Each face shield kit includes one visor and two face shields. Additional visor colors and larger sizes are available. Contact your Avantor sales representative for more information.