Fire Blanket, Sellstrom®
Supplier: Sellstrom
S97450
33000-159EA 406.55 USD
33000-159
Fire Blanket, Sellstrom®
First Aid Supplies Fire and Emergency Blankets
Washable, lightweight, woven fiberglass emergency fire blanket is designed to prevent further fire accidents in the lab.
  • Sparks, flash fires, and flying particles can be easily extinguished
  • Adequately sized to wrap a fire victim
  • To clean, wash with soft sponge and air dry only
  • Sewn-on grommets allow for hanging during cleaning process
Complete with red herculite pouch for wall hanging. Velcro® pull tab on bottom releases 1.5×1.8 m (5×6') blanket.