Anti-CD273 Mouse Monoclonal Antibody (Biotin) [clone: 24F.10C12]

Supplier: Biolegend
329604
329604-BLEA 237.6 USD
329604-BL
Anti-CD273 Mouse Monoclonal Antibody (Biotin) [clone: 24F.10C12]
Antibodies
Biotin anti-human CD273 (B7-DC, PD-L2) [24F.10C12]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD273
Clonality: Monoclonal
Clone: 24F.10C12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR