Anti-CD273 Mouse Monoclonal Antibody [clone: 24F.10C12]

Supplier: Biolegend
329602
329602-BLEA 124.19 USD
329602-BL
Anti-CD273 Mouse Monoclonal Antibody [clone: 24F.10C12]
Antibodies
Purified anti-human CD273 (B7-DC, PD-L2) [24F.10C12]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood.

Type: Primary
Antigen: CD273
Clonality: Monoclonal
Clone: 24F.10C12
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR