Hand-Specific Glove Dispensers, S-Curve Technologies
Supplier: S-Curve Technologies
HS-200-C
32948-896EA 364.97 USD
32948-896
Hand-Specific Glove Dispensers, S-Curve Technologies
Glove Dispensers and Glove Box Holders
Designed to keep gloves clean and protect gloves from ozone degradation.
Ready-to-use glove dispensers feature easily accessible ports that are convenient to fill and maintain. Depth of 38.1cm (15") accommodates long gloves. Dispensers separate right- and left-handed gloves. Constructed of durable 6mm (1/4") thick acrylic.