M006 Nylon Glove Liner, Wells Lamont
Supplier: Wells Lamont
M006M.WLC
32935-568BG 48.32 USD
32935-568
M006 Nylon Glove Liner, Wells Lamont
Gloves
This reusable half-finger liner contains 100% continuous nylon filament.
  • Ambidextrous
  • Reusable
  • For critical environments
  • Washing machine friendly up to 30 times without shrinking
It can be washed 30 times without shrinking, reducing cost of replacement and waste disposal.
Ordering information: These gloves are sold by the dozen pairs.