One-Size-Fits-All Nylon Glove Liners, Wells Lamont
Supplier: Wells Lamont
M088 M089
32935-480CS 479.42 USD
32935-480 32935-536
One-Size-Fits-All Nylon Glove Liners, Wells Lamont
Gloves
Knitted construction provides a one-size-fits-all option to help reduce on-hand glove inventories.
  • Knitted construction
  • Ambidextrous
  • Half-finger and full-finger styles available
  • Full-finger liner also works as gowning or inspection glove
Stretch nylon has limited reuse.
The full-finger liner may also be used as a gowning or inspection glove.
The ambidextrous gloves are available in half-finger and full-finger styles.
Ordering information: Packaged in poly bags of 12 pair.