Whizard® Handguard II White Gloves, Wells Lamont
Supplier: Wells Lamont
333021 333023 333019 333027
32935-139EA 50.9 USD
32935-139 32935-141 32935-143 32935-179 32935-183 87000-551
Whizard® Handguard II White Gloves, Wells Lamont
Gloves
These heavy-duty, cut-resistant gloves feature Honeywell Spectra® Fiber.
  • Prewashed and Preshrunk
  • Honeywell Spectra Fiber
  • Cut Resistant
The combination of high-strength fibers and stainless steel provides exceptional protection against cuts and slashes.
They work best in meat, poultry, and seafood processing.
The prewashed, preshrunk, and ambidextrous gloves have a 3" cuff for wrist protection.
Certifications: ANSI Cut Level 5. CFR 21 approved for food contact.
Ordering information: The gloves are sold in multiples of six pieces.
Caution: Caution Information: Cut-resistant gloves are extremely cut resistant but not cut proof. Do not subject to high speed or highly serrated blades. Always disconnect power before cleaning or removing slicer blades.