PFE Latex Examination Gloves, Kimberly-Clark
Supplier: Kimberly-Clark
57330
32934-007CS 528.1 USD
32934-007 32934-011 32934-015 32934-003 32934-019
PFE Latex Examination Gloves, Kimberly-Clark
Gloves
Gloves provide protection from infectious fluids and bloodborne pathogens.
  • Ambidextrous
  • Beaded cuff
  • For single use only
  • AQL 1.5
  • Thickness: 6.7 mil (finger)
Fully textured finish provides excellent grip in both wet and dry situations. Gloves are powder-free to minimize powder-associated transfer of latex proteins and chemical allergens.
Caution: This product contains natural rubber latex, which may cause allergic reactions. Safe use of this glove by or on latex-sensitive individuals has not been established. This product should not be worn by or exposed to individuals allergic to natural rubber latex.