NeoPro® EC Powder-Free Chloroprene Gloves, Microflex®, Ansell
Supplier: Ansell Healthcare
NEC-288-S NEC-288-XL NEC-288-M NEC-288-L NEC-288-XXL
32916-690CS 386.65 USD
32916-690 32916-696 32916-692 32916-694 32916-698
NeoPro® EC Powder-Free Chloroprene Gloves, Microflex®, Ansell
Gloves
Gloves are made entirely from a synthetic material with comfort close to latex that provides unique protection against a variety of chemicals.
  • Synthetic material with comfort close to latex, provides unique protection against variety of chemicals
  • Exceptional film consistency, manual dexterity, tactile sensitivity, reduced hand fatigue
  • Extended beaded cuff, ideal for high-risk environments
  • Textured fingertips for secure grip
  • Thickness: 8.3 mil (finger)
They offer exceptional film consistency, manual dexterity, tactile sensitivity, fine motor skill function, and reduced hand fatigue. Extended, beaded cuff guards against spills and splashes. Ideal for high-risk environments. Textured fingertips for a more secure grip.
Certifications: Meet ASTM D 6319-99 standards. Approved for use with chemotherapy drugs.
Packaging: Packaged 50 gloves per dispenser box.
Caution: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.