Diamond Grip™ Latex Gloves, Microflex®, Ansell
Supplier: Ansell Healthcare
MF-300-M MF-300-L MF-300-S MF-300-XS MF-300-XL
32916-500CS 304.39 USD
32916-500 32916-502 32916-506 32916-498 32916-503
Diamond Grip™ Latex Gloves, Microflex®, Ansell
Gloves
Thick latex gloves are textured for superior grip and provide excellent protection from rips and tears.
  • Thick textured fingers for gripping in wet and dry conditions
  • Powder-free
  • Fully chlorinated
  • Beaded cuff
  • Thickness: 9.1 mil (finger)
These gloves are thicker than typical latex gloves for enhanced protection. They are thickest at the fingertips, providing superior protection from rips and tears.
Caution: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.