AlphaTec® 56-102 Aprons, Ansell
Supplier: Ansell Healthcare
950104
32899-009CS 217.68 USD
32899-009
AlphaTec® 56-102 Aprons, Ansell
Workwear
Designed for use in food handling and preparation, industrial maintenance, and chemical processing, this versatile CPP™ brand apron offers medium- to heavy-duty splash protection.
  • Wherever oil, grease and chemicals are present, Ansell aprons help workers keep clean and protected from harmful contact
  • PVC (Vinyl) aprons traditionally offer excellent protection where splashes from acids and caustic bases are a concern
  • PVC construction is flexible yet offers resistance to abrasion and puncture
  • Sealed grommets with adjustable ties allow for size adjustment without tear-out
  • Stomach patch enhances protection in critical wear areas
  • Bib styles in two sizes with adjustable ties ensure optimal fit for a wide range of users
Die-cut from virgin vinyl, apron style 56-102 provides superior flexibility and resistance to chemicals, fats, oils, grease, punctures, and abrasions.