Chem Master Neoprene/Rubber Gloves, Showa
Supplier: Showa
CHMM-08 CHMS-07 CHML-09 CHMXL-10
32892-058CS 1079.29 USD
32892-058 32892-057 32892-059 32892-060
Chem Master Neoprene/Rubber Gloves, Showa
Gloves
Hand and product protection for manufacturing, electronics, machine work, food processing, pharmaceutical, and laboratory applications.
  • Resists a broad range of chemicals in many applications
  • 100% cotton flock lining
  • Provide comfort, flexibility, and tactility
  • Self-flushing tractor tread grip for fluid runoff increases gripping effectiveness
Unsupported neoprene over natural rubber construction offers abrasion, tear, and puncture resistantance.
Color: black over blue.
Certifications: Sanitized gloves are made using only listed materials and levels outlined in 21 CFR 170-199.