SHOWA 7714R PVC, Chemical Resistant, Rough Grip, Industrial Glove, Showa
Supplier: Showa
7714R-10
32892-023CS 828.48 USD
32892-023
SHOWA 7714R PVC, Chemical Resistant, Rough Grip, Industrial Glove, Showa
Gloves
PVC gloves provide protection from chemicals and abrasion.
  • Fully coated PVC
  • Excellent abrasion resistance
These gloves are used to handle chemicals. They are cotton lined for extra protection and comfort. Rough grip provides additional grip.
Certifications: ANSI Abrasion Level 4.