SHOWA 7710R PVC, Chemical Resistant, Rough Grip, Industrial Glove, Showa
Supplier: Showa
7710R-10
32892-021CS 673.89 USD
32892-021
SHOWA 7710R PVC, Chemical Resistant, Rough Grip, Industrial Glove, Showa
Gloves
PVC gloves provide protection from chemicals and abrasion.
  • Fully coated PVC
  • Excellent abrasion resistance
These gloves are used to handle chemicals. They are cotton lined for extra protection and comfort. Rough grip provides additional grip.
Certifications: ANSI Abrasion Level 4.