BEST® Neoprene-Coated Gloves, Showa
Supplier: Showa
5122-10
89092-528CS 1980.66 USD
89092-528 32891-976 32891-970 32891-981
BEST® Neoprene-Coated Gloves, Showa
Gloves
BEST® gloves are for manufacturing, refining, petrochemicals, degreasing, plating, handling oils, acids, caustics, alcohols and solvents.
  • Gloves feature a cotton liner and a smooth grip