N-DEX™ Hand-Specific Powder-Free Nitrile Gloves, Showa
Supplier: Showa
9005PFM
32891-830CS 625.17 USD
32891-830 32891-832 32891-838 32891-834
N-DEX™ Hand-Specific Powder-Free Nitrile Gloves, Showa
Gloves
Gloves deliver hand and product protection in food, pharmaceutical, and laboratory applications.
  • Puncture-resistant 100% nitrile construction
  • Smooth finish
  • Rolled cuff
N-DEX™ gloves resist a broad range of chemicals and provide protection from intermittent contact and splashes. Hand-specific, powder-free gloves offer tactile sensitivity, comfort, flexibility, and dexterity for reduced hand fatigue.
Certifications: Gloves are made using only listed materials and levels outlined in FDA CFR 21 170-199. EMT/EMS NFPA 1999 certification.
Ordering information: Gloves feature global sizing; for assistance in choosing the correct glove size, contact your Avantor sales representative. Gloves are dispensed in boxes.