AlphaTec® 87-155 Natural Rubber Latex Gloves with Cotton Lining, Blue, Ansell
Supplier: Ansell Healthcare
120182
32891-404CS 118.37 USD
32891-404 32891-408
AlphaTec® 87-155 Natural Rubber Latex Gloves with Cotton Lining, Blue, Ansell
Gloves
A perfect combination of strength, sensitivity and moderate chemical protection, these natural latex gloves provide a high degree of tensile strength, while the cotton flock lining helps maintain excellent sensitivity.
  • Gloves provide snag, puncture, abrasion and cut resistance
  • Fishscale pattern provides grip on wet or dry objects
Use in the automotive and transportation, food handling and processing, janitorial services, pharmaceuticals, sanitation, and general industries. Uses include canning and food processing; janitorial and maintenance work; and plant maintenance.
Certifications: Complies with FDA food handling requirements (21 CFR 177.2600).
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.