AlphaTec® 29-865 Unsupported Gloves, Ansell
Supplier: Ansell Healthcare
116311 116312 116313 116314 116315
32890-052CS 1838.15 USD
32890-052 32890-054 32890-056 32890-058 32890-060
AlphaTec® 29-865 Unsupported Gloves, Ansell
Gloves
Type 29-865 neoprene gloves offer broad-spectrum protection with excellent resistance to a wide range of chemicals, including oils, acids, caustics, and solvents.
  • Hand-specific
  • Straight cuffs
  • Sand patch finish
  • Case-hardened for increased resistance
Flock-lined gloves are case-hardened to increase chemical and abrasion resistance over ordinary neoprene gloves. Gloves are suitable for petrochemicals and refining operations, cleaning and degreasing, and electronics applications.
Certifications: Complies with FDA food handling requirements 21 CFR (USA) and European Directives.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.