ActivArmr® 27-607 Industrial gloves, oil repellent, Ansell
Supplier: Ansell Healthcare
207310
32889-900CS 2089.62 USD
32889-900
ActivArmr® 27-607 Industrial gloves, oil repellent, Ansell
Gloves
These nitrile gloves are three times more durable than heavy-duty leather, made extra tough for heavy-duty jobs involving rough, abrasive materials.
  • Silicone-free
  • Superior resistance to abrasion
  • More flexible than PVC
  • Excellent barrier to grease and oil
Featuring a unique Ansell nitrile formulation, gloves offer superior resistance to sharp and abrasive materials.
Use in the automotive and transportation, building and construction, machinery and equipment, and metal fabrication industries. Uses include cold plastic mouldings; construction materials, concrete, bricks and tiles; heavy-duty cables; rough castings and cores; rough timber, dry walling and plywood; steel bars, sheet metals, cans and other metal products; and street cleaning and refuse collection.
Stronger and more flexible than PVC, gloves also provide an excellent barrier to grease and oil and do not degrade like leather or cotton. Made without silicone. Guaranteed clean for metals and motor manufacture.
Certifications: ANSI level 1 cut protection; ANSI Level 4 Abrasion protection.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.