Glove Guard® Belt Loop Glove Clip with Safety Breakaway, Magid
Supplier: Magid Glove
GG1939Y
32889-685EA 13.23 USD
32889-685
Glove Guard® Belt Loop Glove Clip with Safety Breakaway, Magid
Gloves
The glove clip with safety breakaway keep gloves readily accessible with glove guards glove clip glove fastener.
  • Clips onto belt loop, heavy waist band, or certain jacket pockets
  • Dielectric
  • Safety break-away capability
The glove guards patented safety breakaway helps prevent individuals from being pulled into machinery, and from being pulled off balance if gloves snag while climbing a ladder.