PolyTuff® 90G Superior Solvent Gloves, QRP
Supplier: PIP
90G
32889-015PK 800.56 USD
32889-015
PolyTuff® 90G Superior Solvent Gloves, QRP
Gloves
The QRP 90G gloves are designed for industrial use with silicon-based adhesives and harsh solvents such as DMF, THF, cyclohexanone, and ketones.
  • Packaged for ISO 5 (Class 100) applications
  • Silicon elastomer formulation withstands flaking, puncture an abrasion
Applications: Silicone based adhesives, harsh solvents microelectronics, avionics, specialty pharmaceutical industries.
Biologically inert PolyTuff silicon elastomer formulation (polysiloxane) withstands flaking, puncture and abrasion. Seamless comfort curve paired RH/LH gloves minimize hand and finger fatigue. ISO 5 (Class 100) processed and packaged.