Butyl Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
B324/8 B131R/10 B131R/8 B131R/9 B131/10 B131/11 B161/10 B174R/8 B324/9 B131/8 B161R/11 B161R/7 B161/7 B161/8 B161R/9 B161/9 B161/11 B174R/10 B174/10 B174/11 B324R/11 B131R/11 B131R/7 B131/9 B174R/11 B174R/9 B324R/8 B131/7 B161R/10 B161R/8 B324R/10 B324R/9
32887-949PR 116.38 USD
32887-949 32887-970 32887-904 32887-910 32887-912 32887-914 32887-916 32887-921 32887-942 32887-960 32887-972 32887-920 32887-926 32887-928 32887-929 32887-931 32887-932 32887-935 32887-936 32887-938 32887-946 32887-948 32887-954 32887-906 32887-908 32887-922 32887-940 32887-944 32887-950 32887-956 32887-918 32887-924 32887-930 32887-952 32887-958 32887-962
Butyl Gloves, Honeywell Safety
Butyl gloves exhibit high permeation resistance to gas and water vapors.
  • High permeation resistance to gas and water vapors
  • Resistant to oxygenated solvents and most oxidizing chemicals
  • Remain flexible and sensitive, even at lower temperatures
  • Hand specific, curved design improves fit and comfort
  • Rolled cuff
Gloves stay flexible and sensitive, even at lower temperatures. Curved finger and hand design provide a comfortable fit. Gloves with a rough finish are excellent for wet applications. Rolled cuff. Hand-specific.
Resistant to oxygenated solvents and most oxidizing chemicals.