Butyl Gloves
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
Exhibit high permeation resistance to gas or water vapors. Ideal for use in ketones and esters. Flexible and sensitive, even at lower temperatures. Curved fingers and hand design. Thickness: 16 mil. Rough finish. Size 9.