SP Bel-Art Clavies™ Biohazard Autoclave Gloves, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H13201-0000 H13201-0001 H13201-0000
32885-805CS 748.49 USD
32885-805 32885-826 490012-180
SP Bel-Art Clavies™ Biohazard Autoclave Gloves, Bel-Art Products, a part of SP
Gloves
Soft, pliant, all-cotton terry cloth gloves are superb for handling hot objects out of the autoclave or oven, or for other heat protection in the lab, school or industry.
  • Heat-resistant
  • Machine wash, air dry
Heat-resistant up to 232 °C (450 °F), they replace hazardous asbestos gloves. Gauntlets in two lengths provide wrist and forearm protection, and the bright orange color offers high visibility.
Caution: Gloves are not waterproof and should not be used to handle dry ice.