Qualatherm® 75G Mid-Temperature Cleannroom Gloves, 23", QRP
Supplier: PIP
75G
32885-785PK 2355.15 USD
32885-785
Qualatherm® 75G Mid-Temperature Cleannroom Gloves, 23", QRP
Gloves
The QRP 75G gloves are RoHS compliant gloves. PolyTuff silicon elastomer formulation offers clean, liquid-proof mid-temperature protection for wet or dry environments.
  • Packaged for ISO 5 (Class 100)
  • Wet or dry contact temperatures from –78 to 450 °F (–61.4 to 232 °C)
  • PolyTuff silicone elastomer is sulfur-free
  • Excellent resistance to chemicals, solder and fluxes
Applications of the QRP 70G Qualatherm 450 °F mid-temperature cleanroom gloves include: Diffusion furnaces, Quartz wafer boats, Burn-in ovens, PWB handling after wave soldering, ceramics, porcelains, food processing and handling.
PolyTuff silicone elastomer formulation offers clean, liquid-proof mid-temperature protection for wet or dry environments. Excellent resistance to chemicals, solders and fluxes, remaining flexible even at elevated temperatures. Seamless Comfort Curve™ paired Right Hand/Left Hand gloves minimize hand and finger fatigue. Free from PCB, asbestos and fiberglass hazards; Latex-Free. Non-particulating, cut and abrasion resistant. Micro-coated, high density, continuous filament nylon liner assures clean, easy donning and removal.