SP Bel-Art Beta Locking Storage Box, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F24987-0000
32800-080EA 657.15 USD
32800-080
SP Bel-Art Beta Locking Storage Box, Bel-Art Products, a part of SP
Boxes Beta/Gamma Radiation Boxes
For secure storage of radioactive materials, such as 32P, 35S and 14C.
  • Clear acrylic construction
  • Hinged lid
  • Easy to secure with anti-theft connectors
Fabricated from 9.5 mm (3/8") thick clear acrylic for all-around visibility of stored contents. A hinged lid with hasp is provided for padlocking. Entire unit may be secured with use of a locking cable through the anti-theft connectors securely attached to the box. Lock and cable not included.
Certifications: Complies with USNRC, Title 10, Chapter 1, Part 20.