PYREX® 75° Angle Three-Way Connecting Adapter, Standard Taper Joints
Supplier: Corning
9000-19 9000-24 9000-14
32645-356CS 125.8 USD
32645-356 89091-662 62949-961
PYREX® 75° Angle Three-Way Connecting Adapter, Standard Taper Joints
Adapters Adapters, Conical Ground Joints
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These angle connecting adapters have a sidearm at an approximately 75° angle from the lower joint. The lower end and sidearm are equipped with inner 24/40 standard taper joints, and the upper end with an outer 24/40 standard taper joint.