Dextrose 5% USP for injection

Supplier: B BRAUN MEDICAL

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

L5100
32500-203CS 632.11 USD
32500-203
Dextrose 5% USP for injection
D(+)-Glucose

Dextrose Injections USP are sterile, nonpyrogenic and contain no bacteriostatic or antimicrobial agents. These products are intended for intravenous administration.


These intravenous solutions are indicated for use in adults and pediatric patients as sources of calories and water for hydration.


Not made with latex, PVC or DEHP.


Specific Gravity: 1.017


NDC number: 0264-7510-00

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146
MDL Number: MFCD00148912
CAS Number: 50-99-7
Merck Index: 14,04459

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR