KIMAX® KIMBLE® Gas Measuring Tubes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
30060-50
32375-025EA 182.37 USD
32375-025
KIMAX® KIMBLE® Gas Measuring Tubes, DWK Life Sciences
Tubes Gas Sampling Tubes
KIMAX® gas measuring tube is closed at the zero end for gas measurement.
  • Graduation interval: 0.1 ml
  • Graduation range: 0.5 to 50 ml
  • Tolerance: ±0.10 ml
Manufactured from precision bore tubing. Durable black ceramic enamel scale without stopcock.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.