Anti-DR3 (TRAMP) Mouse Monoclonal Antibody (Biotin) [clone: JD3]

Supplier: Biolegend
307104
307104-BLEA 298.3 USD
307104-BL
Anti-DR3 (TRAMP) Mouse Monoclonal Antibody (Biotin) [clone: JD3]
Antibodies
Biotin anti-human DR3 (TRAMP) [JD3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: DR3 (TRAMP)
Clonality: Monoclonal
Clone: JD3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR