Pestle Motor for Pellet Pestel Mixers, Cordless, Chemglass
Supplier: Chemglass
CLS-5001-001
30624-128EA 228.81 USD
30624-128
Pestle Motor for Pellet Pestel Mixers, Cordless, Chemglass
Mortars and Pestles Pestles
The Cordless pestle motor for pellet pestle mixers are light-weight, handheld, battery-powered motor designed to drive disposable polypropylene pestles in 0.5 and 1.5 mL microcentrifuge tubes
  • Tip adapter securely holds the pestles while in operation
  • A new polypropylene pestle is used with each sample
  • DNase, RNase and Pyrogen-Free
  • Autoclavable
Protein and DNA pellets, precipitates, soft tissues and other solids are completely dispersed or resuspended in seconds by a combination of mechanical and vortex action.
Delivery information: Includes tip adapter and two AA batteries.