CoolCube™ Microtube and Microplate Cooler, Chemglass
Supplier: Chemglass
CLS-3200-001
30623-436EA 233.38 USD
30623-436
CoolCube™ Microtube and Microplate Cooler, Chemglass
Coolers Benchtop Coolers
The CoolCube™ is designed to store samples safely on the lab bench without sample degradation due to temperature increase or fluctuation.
  • Constructed of DuraStar (a high strength polymer) and a patented gel refrigerant
  • Maintains temperature at approximately 0°C for up to four hours
One side of the CoolCube™ cooler can be used as a convenient PCR workstation with wells for 12 standard 1.5/2.0 mL microtubes and one 96 well PCR plate or 0.2 mL tubes/strips. The other side can be used to hold 36 standard 1.5/2.0 mL microtubes.