BioClave™ Benchtop Autoclaves, Chemglass
Supplier: Chemglass
CLS-1191-008
30622-734EA 7261.49 USD
30622-734 30622-736
BioClave™ Benchtop Autoclaves, Chemglass
Sterilizers Autoclaves (Steam Sterilizers)
Steam sterilization is convenient and affordable with the new BioClave™ digital research autoclaves.
  • It includes a mechanical and electrical safety interlock
  • It prevents the door from being opened until the pressure has reached zero psi
  • With a width of less than 18 inches it will fit comfortably on even the most crowded bench tops
  • Three trays are supplied with each 16L unit (two for 8L unit) along with a convenient tray removal tool
The sterilization chambers are large, accommodating a variety of liquids, media, instruments, glassware, plastic ware and other common laboratory items
The 16 liter BioClave™ offers three cycle options (standard programs), which are preset for the sterilization of liquids, wrapped instruments or unwrapped instruments/plastic ware. A fourth option, “Dry Only”, is also available for additional drying tie to be added to the end of a cycle. For special applications requiring variations in cycle parameters, all settings on the 16 liter BioClave™ are fully adjustable.
The 8 liter BioClave™ Mini is preprogrammed to meet basic sterilization needs. Although the temperature can be set at either 121°C or 134°C, the sterilization time is fixed. The economical BioClave™ Mini is the ideal choice wherever bench top sterilization is required and space is limited.
With large digital displays and fully automatic operation, all segments of the sterilization cycle (fill, sterilize, exhaust and dry) commence and run to completion by simply pressing the start button.
Certifications: CE approved.