Clamp Holders, Chemglass
Supplier: Chemglass
CG-2040-C-01
30622-652EA 65.76 USD
30622-652
Clamp Holders, Chemglass
Clamps General Purpose Clamps
Heavy duty epoxy coated alloy clamp has high tensile strength and corrosion resistance
Can be used to support stirrers, temperature monitors and condensers. Large hand knobs allow easy adjusment and secure clamping power. Useful wherever clamping at 90° is required.