Straight Side Glass Bottles, Amber, C&G Containers
Supplier: C&G Containers
LAS004200020 LAS004900000 LAS009200010 LAS009900000 LAS0082000 LAS008100010 LAS009100000 LAS009200000 LAS004100000
71000-686CS 121.63 USD
71000-686 71000-690 71000-692 71000-694 71000-696 89125-426 89125-428 89137-136 30621-180 30621-182 30621-184 30621-186
Straight Side Glass Bottles, Amber, C&G Containers
Bottles Glass Bottles
These glass jars are used primarily for the collection of liquid media, soil, sediments, and slurry media for testing Semi-volatiles (light sensitive compounds), pesticides, PCBs, diesel range organics, TPH, metals, and phenolics.
Pre-cleaned and certified products contain the certificate of analysis and cleaning protocol form in the product case.
A lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement for pre-cleaned and certified products. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (Nitric acid wash), B1 (Deionized water wash), C1 (Hydrochloric acid wash), and L3 (no wash). Some bottles are tare weighed.
Secondary uses include containment, bench vessels, and archiving. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity 'contain' volumes may vary.