RotoFlex Plus Tube Rotator, Argos Technologies
Supplier: Antyila Scientific
04397-40
30621-084EA 1299.48 USD
30621-084
RotoFlex Plus Tube Rotator, Argos Technologies
Shakers and Mixers Rotating/Rocking Shakers
The RotoFlex Plus is a variable speed, digitally controlled tube rotator ideal for DNA Purification, protein binding, hybridization, and sample mixing
The RotoFlex Plus can also be used with a magnetic platform (sold separately) to optimize Western Blot hybridization. Simply attach your plastic bag containing membrane and antibody solution to the plate using the metal strips provided.
It will accommodate tubes from 0.5 – 50mL by using included rotisserie panels. The easy to operate key pad allows for speed adjustments between 10 – 40 rpm. In addition to the variable speed feature the unit can also be operated in an oscillating mode. In oscillation mode the rotisseries rotate 40°, pause, then oscillate back and forth five times before rotating another 40° to repeat the process. After oscillating for 360°, the rotisserie rotates a complete turn before reverting back to the oscillation cycle.
Ordering information: Unit is supplied with base unit and rotisseries (one pack of each size).