Stir Vane, PTFE, Dynalon
Supplier: Dynalon
304465-0002
30620-500CS 177.79 USD
30620-500
Stir Vane, PTFE, Dynalon
Stirrers Magnetic Stirring Bars
Solid PTFE resin vanes feature molded magnetic bars in center
Vanes permit rotation in small conical tubes, and ensure thorough sample mixing. PTFE offers excellent chemical resistance. Always store in pairs to maintain magnetic strength.