Stir Bar Retriever, Polypropylene, Dynalon
Supplier: Dynalon
303325
30620-472CS 185.16 USD
30620-472
Stir Bar Retriever, Polypropylene, Dynalon
Stirrers Stirring Tools
The polypropylene stir bar retriever can be used with all stir bars
Polypropylene is chemically resistant and may be used up to 135 °C.
Delivery information: Includes hanging ring for storage convenience.
Packaging: Weight: 0.05375 lbs.