Buchner Funnel Discs, Dynalon
Supplier: Dynalon
242845-110
30620-312CS 147.33 USD
30620-312
Buchner Funnel Discs, Dynalon
Filters Filter Discs
Made from high density polyethylene, these Buchner Funnel discs offer excellent chemical resistance.
They are useful for holding up filter paper, ensuring that blocking of the holes is avoided and the filtering surface of the funnel is free. Discs will withstand temperatures up to 110 °C (230 °F). Mesh size is 950×700 µm.