Container with Hinged Lid, Dynalon
Supplier: Dynalon
226254-2000
30620-252CS 233.95 USD
30620-252
Container with Hinged Lid, Dynalon
Sample Containers General Purpose Containers
These hinged lid containers are ideal for use virtually anywhere
They feature a secure snap closure with hinged lid that eliminates lost caps. Containers offer air and moisture resistance to contents. Manufactured from flexible FDA approvable polyethylene resin.