Refractive Index Liquids, Series E
Supplier: CARGILLE LABORATORIES INC.
1843Y
30617-960EA 477.75 USD
30617-960
Refractive Index Liquids, Series E
Standards Refractive Index Standards
Refractive index liquids have a broad range of applications in diversified fields.
  • Slightly volatile
  • Colorless flurocarbon formulation
Adjustment: ±0.0002
Choose from Range nD 1.500 to 1.640.with liquids at 0.0005 intervals.
Ordering information: Required refractive index has to be specified with order.