PFA Vessel, 2.4 L, Savillex
Supplier: SAVILLEX
700-960
30617-548EA 1248.56 USD
30617-548
PFA Vessel, 2.4 L, Savillex
Sample Containers General Purpose Containers
This 15.2cm (6") diameter PFA vessel has a capacity of 2.4L and is ideal for cleaning ICP glassware, along with many other uses
Service temperature range is –200 to 260°C (–328 to 500°F).
Manufactured from the highest purity PFA resin for ultratrace metals use. Flat bottomed for hotplate use. Flanged rim allows for easy handling. PFA cover sold separately.
Ordering information: PFA cover with handle sold separately.