PFA Pipette Tips, Savillex
Supplier: SAVILLEX
700-600
30617-536EA 57.11 USD
30617-536
PFA Pipette Tips, Savillex
Pipette Tips
These PFA pipette tips are made from the highest purity grade PFA resin and are designed for the most demanding ultratrace metals analysis applications
For a constant supply of high purity acid, use the Savillex DST-1000 Acid Purification System (30621-038).
The 8mm opening fits most popular 1000µL and adjustable pipettes. Orifice size is 8mm. Designed to be reused after cleaning in ultrapure acid.