Wash Bottle, PFA, Savillex
Supplier: SAVILLEX
700-500
30617-534EA 653.72 USD
30617-534
Wash Bottle, PFA, Savillex
Bottles Wash Bottles
This 500 ml PFA wash bottle is manufactured using Savillex's proprietary molding technology, which produces an extremely smooth surface.
The spout, closure, and bottle are all made from the highest purity PFA resin, making the wash bottle suitable for the most demanding ultratrace applications. Ideal for use with ICP-MS.