Qualitative Filter Paper, Grade 609, Ahlstrom
Supplier: Ahlstrom
6090-3300
30617-436PK 153.11 USD
30617-436
Qualitative Filter Paper, Grade 609, Ahlstrom
Filters Filter Paper Qualitative Filter Paper
Ahlstrom Grade 609 is an evolution of Ahlstrom 613 for use in the beer industry.
This is a filter paper with an extremely low iron and nitrogen content for the analysis of protein in beer.