Wood Funnel Support, Bennett Wood
Supplier: Bennett Wood Specialties
2635
30580-001EA 411.12 USD
30580-001
Wood Funnel Support, Bennett Wood
Support Stands
Designed to hold six separatory funnels
With adjustable upper arm and auxiliary middle arm similarly slotted, and adjustable in height. Without glassware.